Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.01800

Sigma-Aldrich

Benzonitrile

for synthesis

Synonym(s):

Benzonitrile, Phenyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5N
CAS Number:
Molecular Weight:
103.12
MDL number:
UNSPSC Code:
12352117
EC Index Number:
202-855-7
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.7 hPa ( 20 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

autoignition temp.

550 °C (experimental)

potency

690 mg/kg LD50, oral (Rat)

expl. lim.

1.4-7.2 % (v/v)

bp

190 °C/1013 hPa

mp

-13 °C

transition temp

flash point 70 °C

solubility

10 g/L

density

1.00 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N#Cc1ccccc1

InChI

1S/C7H5N/c8-6-7-4-2-1-3-5-7/h1-5H

InChI key

JFDZBHWFFUWGJE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service