Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.01658

Sigma-Aldrich

2-Bromoacetophenone

for synthesis

Synonym(s):

2-Bromoacetophenone, Phenacyl bromide, ω-Bromoacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7BrO
CAS Number:
Molecular Weight:
199.04
MDL number:
UNSPSC Code:
12352115
EC Index Number:
200-724-9
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (GC)

form

crystals

bp

140 °C/15 hPa

mp

48-51 °C

transition temp

flash point >112 °C

density

1.65 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H7BrO/c9-6-8(10)7-4-2-1-3-5-7/h1-5H,6H2

InChI key

LIGACIXOYTUXAW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 47 °C
Melting range (upper value): ≤ 50 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service