Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.01610

Sigma-Aldrich

1,4-Bis(hydroxymethyl)-cyclohexane

(mixture of cis- and trans-isomers) for synthesis

Synonym(s):

1,4-Bis(hydroxymethyl)-cyclohexane, 1,4-Cyclohexylenedimethanol, 1,4-Dimethylolcyclohexane, 1,4-Cyclohexanedimethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16O2
CAS Number:
Molecular Weight:
144.21
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-268-9
NACRES:
NA.22

vapor pressure

<0.24 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

autoignition temp.

330 °C (DIN 51794)

potency

>2000 mg/kg LD50, oral (Rat)

pH

4.2 (20 °C, 250 g/L in H2O)

bp

286 °C/1013 hPa

mp

34.6 °C

transition temp

flash point 160 °C (DIN 51758)

density

1.04 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OCC1CCC(CC1)CO

InChI

1S/C8H16O2/c9-5-7-1-2-8(6-10)4-3-7/h7-10H,1-6H2

InChI key

YIMQCDZDWXUDCA-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

321.8 °F - closed cup

Flash Point(C)

161 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service