Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.01299

Sigma-Aldrich

1,3,5-Benzenetricarboxylic acid

for synthesis

Synonym(s):

1,3,5-Benzenetricarboxylic acid, Trimesic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3-1,3,5-(COOH)3
CAS Number:
Molecular Weight:
210.14
MDL number:
UNSPSC Code:
12352106
EC Index Number:
209-077-7

Assay

≥95.0% (HPLC)

autoignition temp.

590 °C

potency

16000 mg/kg LD50, oral (Rat)

mp

>300 °C

transition temp

flash point 328 °C

solubility

26.3 g/L

bulk density

720 kg/m3

InChI

1S/C9H6O6/c10-7(11)4-1-5(8(12)13)3-6(2-4)9(14)15/h1-3H,(H,10,11)(H,12,13)(H,14,15)

InChI key

QMKYBPDZANOJGF-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 95.0 % (a/a)
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service