Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.01228

Sigma-Aldrich

11-Aminoundecanoic acid

for synthesis

Synonym(s):

11-Aminoundecanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$234.00

$234.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$234.00

About This Item

Linear Formula:
H2N(CH2)10COOH
CAS Number:
Molecular Weight:
201.31
MDL number:
UNSPSC Code:
12352106
EC Index Number:
219-417-6
NACRES:
NA.22

$234.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

powder

mp

183-187 °C

bulk density

500 kg/m3

storage temp.

2-30°C

SMILES string

[N+H3]CCCCCCCCCCC(=O)[O-]

InChI

1S/C11H23NO2/c12-10-8-6-4-2-1-3-5-7-9-11(13)14/h1-10,12H2,(H,13,14)

InChI key

GUOSQNAUYHMCRU-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Melting range (lower value): ≥ 183 °C
Melting range (upper value): ≤ 187 °C
Identity (IR): passes test

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service