Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.00898

Sigma-Aldrich

Diethyl malonate

for synthesis

Synonym(s):

Diethyl malonate, Ethyl malonate, Malonic acid diethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$57.00
500 ML
$66.70
1 L
$102.00
2.5 L
$174.00

$57.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$57.00
500 ML
$66.70
1 L
$102.00
2.5 L
$174.00

About This Item

Linear Formula:
CH2(COOC2H5)2
CAS Number:
Molecular Weight:
160.17
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-305-9
NACRES:
NA.22

$57.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

1.3 hPa ( 40 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

435 °C (DIN 51794)

potency

15720 mg/kg LD50, oral (Rat)
>16000 mg/kg LD50, skin (Rabbit)

expl. lim.

0.8-12.8 % (v/v)

bp

199 °C/1013 hPa

mp

-50 °C

transition temp

flash point 90 °C (DIN 51758)

solubility

20.8 g/L (External MSDS)

density

1.055 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CC)C(=O)CC(=O)OCC

InChI

1S/C7H12O4/c1-3-10-6(8)5-7(9)11-4-2/h3-5H2,1-2H3

InChI key

IYXGSMUGOJNHAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.053 - 1.057
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service