Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.00871

Sigma-Aldrich

Bromoethane

for synthesis

Synonym(s):

Bromoethane, Ethyl bromide

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$39.80
1 L
$117.00

$39.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$39.80
1 L
$117.00

About This Item

Empirical Formula (Hill Notation):
C2H5Br
CAS Number:
Molecular Weight:
108.97
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-825-8
NACRES:
NA.22

$39.80


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

510 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

510 °C

potency

1350 mg/kg LD50, oral (Rat)

expl. lim.

6.7-11.3 % (v/v)

bp

37-39 °C/1013 hPa

mp

-118 °C

transition temp

flash point -20 °C

solubility

9 g/L

density

1.46 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

BrCC

InChI

1S/C2H5Br/c1-2-3/h2H2,1H3

InChI key

RDHPKYGYEGBMSE-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.458 - 1.462
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Ozone 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-9.4 °F - closed cup

Flash Point(C)

-23 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service