Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.00826

Sigma-Aldrich

Acryloyl chloride

(stabilised with phenothiazine) for synthesis

Synonym(s):

Acryloyl chloride, Acrylic acid chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$141.00
500 ML
$499.00

$141.00


Please contact Customer Service for Availability


Select a Size

Change View
100 ML
$141.00
500 ML
$499.00

About This Item

Linear Formula:
CH2=CHCOCl
CAS Number:
Molecular Weight:
90.51
MDL number:
UNSPSC Code:
12352106
EC Index Number:
212-399-0

$141.00


Please contact Customer Service for Availability

vapor pressure

106.6 hPa ( 20 °C)

Quality Level

Assay

≥96.0% (GC)

bp

74-76 °C/1013 hPa

transition temp

flash point -4 °C

density

1.113-1.119 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

ClC(=O)C=C

InChI

1S/C3H3ClO/c1-2-3(4)5/h2H,1H2

InChI key

HFBMWMNUJJDEQZ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.112 - 1.119
Identity (IR): passes test
The material is stabilised with Phenothiazine. At time of analytical testing a content of >100 ppm stabiliser was determined.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

30.2 °F

Flash Point(C)

-1 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service