Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.00239

Sigma-Aldrich

Benzyl cinnamate

for synthesis

Synonym(s):

Benzyl cinnamate, Benzyl trans-3-phenylpropenoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14O2
CAS Number:
Molecular Weight:
238.28
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-109-3

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

crystals

potency

5530 mg/kg LD50, oral (Rat)
>3000 mg/kg LD50, skin (Rabbit)

bp

350 °C/1013 hPa

mp

34 °C

transition temp

flash point 180 °C

solubility

<10 mg/L

density

1.109 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C16H14O2/c17-16(12-11-14-7-3-1-4-8-14)18-13-15-9-5-2-6-10-15/h1-12H,13H2/b12-11+

InChI key

NGHOLYJTSCBCGC-VAWYXSNFSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 33 °C
Melting range (upper value): ≤ 36 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(C)

180 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service