Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.00192

Sigma-Aldrich

Octanoic acid

for synthesis

Synonym(s):

Octanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$45.10
1 L
$96.50
2.5 L
$182.00

$45.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$45.10
1 L
$96.50
2.5 L
$182.00

About This Item

Linear Formula:
CH3(CH2)6COOH
CAS Number:
Molecular Weight:
144.21
MDL number:
UNSPSC Code:
12352106
EC Index Number:
204-677-5
NACRES:
NA.22

$45.10


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

0.05 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

>300 °C

potency

>2000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

expl. lim.

1 % (v/v)

pH

4 (20 °C, 0.2 g/L in H2O)

bp

237 °C/1013 hPa

mp

16 °C

transition temp

flash point 130 °C

solubility

0.68 g/L

density

0.91 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[O-]C(=O)CCCCCCC.[H+]

InChI

1S/C8H16O2/c1-2-3-4-5-6-7-8(9)10/h2-7H2,1H3,(H,9,10)

InChI key

WWZKQHOCKIZLMA-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.908 - 0.912
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service