Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.00187

Sigma-Aldrich

2-Ethoxyethyl acetate

for synthesis

Synonym(s):

2-Ethoxyethyl acetate, Ethylene glycol monoethyl ether acetate, Ethyl glycol acetate, Acetic acid 2-ethoxyethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O3
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-839-2
Assay:
≥99% (GC)
bp:
156 °C/1013 hPa
vapor pressure:
2.66 hPa ( 25 °C)

vapor pressure

2.66 hPa ( 25 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

autoignition temp.

379 °C

potency

10300 mg/kg LD50, skin (Rabbit)

expl. lim.

1.7-10 % (v/v)

pH

4-5 (20 °C in H2O, saturated solution)

bp

156 °C/1013 hPa

mp

-62 °C

transition temp

flash point 52 °C

solubility

247 g/L

density

0.97 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C6H12O3/c1-3-8-4-5-9-6(2)7/h3-5H2,1-2H3

InChI key

SVONRAPFKPVNKG-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.972 - 0.974
Water (K. F.): ≤ 0.10 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service