Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.00184

Sigma-Aldrich

Azodicarboxamide

for synthesis

Synonym(s):

Azodicarboxamide, Azodicarbonamide, 1,1-Azobisformamide, Dicarbamoyldiimide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
116.08
MDL number:
UNSPSC Code:
12352100
EC Index Number:
204-650-8

vapor pressure

<1 Pa ( 25 °C)

Quality Level

form

powder

potency

>2000 mg/kg LD50, oral (Rat)
>500 mg/kg LD50, skin (Rat)

pH

7 ( in H2O, neutral)

mp

>200 °C (decomposition)

transition temp

flash point >200 °C

solubility

0.033 g/L

density

1.61 g/cm3 at 20 °C

storage temp.

15-25°C

InChI

1S/C2H4N4O2/c3-1(7)5-6-2(4)8/h(H2,3,7)(H2,4,8)

InChI key

XOZUGNYVDXMRKW-UHFFFAOYSA-N

General description

for synthesis

Application

Azodicarboxamide for synthesis. CAS No. 123-77-3, EC Number 204-650-8.

Analysis Note

Assay (iodometric)≥ 98.0 %Identity (IR)passes test

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Resp. Sens. 1

Supplementary Hazards

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 1

Flash Point(C)

> 200 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service