Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73294BC

Millipore

mEHEC® 15.8 g Media Scoop

BioControl®, For use with mEHEC®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122419

storage condition

room temperature

application(s)

food and beverages
pathogen testing

compatibility

BioControl, For use with mEHEC

Components

One 15.8 g scoop

Compatibility

BioControl® GDS

Testing Method

PCR

Legal Information

BioControl is a registered trademark of BioControl Systems, Inc.
mEHEC is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service