Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

71752-M

Sigma-Aldrich

LB Agar, Miller - Novagen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

form

solid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

storage temp.

10-30°C

General description

LB Agar MILLER is a granulated medium for the cultivation of <em>E. coli</em>. The granules ensure rapid and uniform dissolution in water, and prevent clumping of the medium and inhalation of the airborne powder. It is supplied in several formats. EasyPak is an aluminum foil pouch containing 37 g granulated medium sufficient for 1 L culture. Just add the EasyPak contents to 1 L water, microwave to dissolve agar and sterilize*, and it is ready to use. In addition, the LB Agar MILLER is supplied in 500 g, and 5 kg bottles. The composition of the LB Agar MILLER per liter is 5 g yeast extract, 10 g peptone from casein, 10 g sodium chloride, and 12 g agar-agar.

*Microwaving is sufficient if an antibiotic will be added to the prepared media.

Warning

Toxicity: Standard Handling (A)

Legal Information

71752-5 and 71752-6 are not available for sale in Austria, Belgium, Denmark, Finland, France, Germany, Ireland, Italy, Luxembourg, Netherlands, Norway, Portugal, Spain, Sweden, Switzerland, and UK.
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service