Skip to Content
MilliporeSigma
All Photos(1)

Documents

71591-M

Sigma-Aldrich

StrepTag II Antibody HRP Conjugate

Novagen®

Synonym(s):

HRP StrepTag II, StrepTag II Detection

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

conjugate

horse radish peroxidase (HRP)

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

form

liquid

manufacturer/tradename

Novagen®

storage condition

OK to freeze

technique(s)

western blot: suitable

storage temp.

2-8°C

General description

The Strep•Tag II Antibody HRP Conjugate for one-step affinity purification of Strep•Tag<TMSYMBOL></TMSYMBOL> fusion proteins.

The Strep•Tag II Antibody HRP Conjugate*is a peroxidase-conjugated Strep•Tag IIMonoclonal Antibody* that is useful as asingle detection reagent thus eliminatingcross-reactivity associated with secondaryreagents.The Strep•TagII Antibody HRP Conjugate can detect as littleas 5 ng Strep•Tag II fusion protein.

* Strep•Tag<TMSYMBOL></TMSYMBOL> technology for protein purification and detection is covered under US Patent No. 5,506,121, UK Patent No. 2,272,698 andFrench Patent No. 93 13 066. Strep•Tactin<TMSYMBOL></TMSYMBOL> is covered by US Patent No. 6,103,493. These products are provided only for research use.Information about licenses for commercial use is available from IBA GmbH, Rudolf-Wissell-Str. 28, D-37079 Goettingen, Germany.

Application

Strep-Tag II Antibody HRP Conjugate produces a strong signal at a 1:4000 dilution for dot blots and western blots.

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
StrepTrap is a trademark of Cytiva

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masatoshi Wakamori et al.
Protein expression and purification, 74(1), 88-98 (2010-07-06)
The bacterial co-expression method is a useful protein expression technique to reconstitute a hetero-oligomeric protein complex in vitro. However, during the plasmid subcloning for co-expression, unintended cleavage at the sequences of target cDNAs becomes more frequent as the number of
Shrijan Bhattarai et al.
Microbiology spectrum, 11(3), e0486122-e0486122 (2023-05-25)
Staphylococcus aureus utilizes the two-component regulatory system VraSR to receive and relay environmental stress signals, and it is implicated in the development of bacterial resistance to several antibiotics through the upregulation of cell wall synthesis. VraS inhibition was shown to
Kazuhiro Kashiwagi et al.
Nature, 531(7592), 122-125 (2016-02-24)
Eukaryotic cells restrict protein synthesis under various stress conditions, by inhibiting the eukaryotic translation initiation factor 2B (eIF2B). eIF2B is the guanine nucleotide exchange factor for eIF2, a heterotrimeric G protein consisting of α-, β- and γ-subunits. eIF2B exchanges GDP
Nadia Martinez-Martin et al.
Cell, 174(5), 1158-1171 (2018-07-31)
Characterizing cell surface receptors mediating viral infection is critical for understanding viral tropism and developing antiviral therapies. Nevertheless, due to challenges associated with detecting protein interactions on the cell surface, the host receptors of many human pathogens remain unknown. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service