Skip to Content
MilliporeSigma
All Photos(1)

Documents

71514-M

Millipore

Floating Rack, Maxi

Synonym(s):

Floating Rack for D-Tube Dialyzers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900
NACRES:
NB.24

manufacturer/tradename

Novagen®

storage condition

OK to freeze

storage temp.

10-30°C

General description

The Floating Rack is designed to hold D-Tube Dialyzers in an exchange buffer. Each Floating Rack, Mini can hold up to nine D-Tube Dialyzer, Mini devices. Each Floating Rack, Midi can hold up to four D-Tube Dialyzer, Midi units. Each Floating Rack, Maxi can hold up to seven D-Tube Dialyzer, Maxi devices. Each Floating Rack, Mega can hold one D-Tube Dialyzer, Mega unit. Each package contains 10 Floating Racks.

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service