Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

71404-M

Sigma-Aldrich

Rosetta 2(DE3)pLacI Competent Cells - Novagen

Escherichia coli, rod shaped

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106202
NACRES:
NA.81

product name

Rosetta 2(DE3)pLacI Competent Cells - Novagen, Rosetta strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli. Contains the pLacI plasmid producing extra Lac repressor.

biological source

Escherichia coli

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

cell transformation

transformation efficiency: 2×106 cfu/μg

shipped in

dry ice

storage temp.

−70°C

General description

Genotype: FompT hsdSB (rB mB) gal dcm (DE3) pLacIRARE2 (CamR)





This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
Expression host strains for pETBlue and pTriEx vectors are lDE3 lysogens that also carry the compatible pLacI plasmid, encoding the lac repressor. The pLacI plasmid confers chloramphenicol resistance and provides enough lac repressor to inhibit transcription from the T7lac promoter in the absence of inducer. The NovaBlue host strain is recommended for initial cloning with pETBlue and pTriEx vectors due to its superior transformation efficiency and high yields of high-quality plasmid DNA.

Note that these strains are not recommended for use with pET, pACYCDuet, pCDF, pRSF, pCOLADuet or pETcoco vectors.

.Rosetta 2 strains are BL21 derivatives designed to alleviate codon bias when expressing heterologous proteins in E. coli. The cells carry a chloramphenicol-resistant plasmid with the tRNA genes that decode seven rare codons (AGA, AGG, AUA, CUA, GGA, CCC, and CGG) to improve the yield of full-length proteins. The tRNA genes are driven by their native promoters. In Rosetta 2(DE3)pLacI, the rare tRNA genes are present on the same plasmid that carries the lac repressor gene.

DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
Rosetta strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli. Contains the pLacI plasmid producing extra Lac repressor.

Components

0.4 ml1 mlComponent

•2 × 0.2 ml5 × 0.2 mlRosetta 2(DE3)pLacI Competent Cells

•2 × 2 ml4 × 2 mlSOC Medium

•2 ng2 ngTest Plasmid

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service