Skip to Content
MilliporeSigma
All Photos(1)

Documents

71279

Sigma-Aldrich

Veggie Yeast Extract - Novagen

Synonym(s):

Veggie Extract

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106212
NACRES:
NA.54

form

solid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

shipped in

ambient

storage temp.

10-30°C

General description

Veggie Yeast Extract is a certified animal-free media component that can be used as a direct replacement for traditional yeast extract in bacterial growth media. It has been quality tested to ensure proper growth and maintenance of bacterial cells. The Veggie line of products are ideal for applications requiring minimal use of animal-derived materials.

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service