Skip to Content
MilliporeSigma
All Photos(1)

Documents

71210-M

Sigma-Aldrich

(DE3)pLysS Competent Cell Set 2 - Novagen

Escherichia coli, rod shaped

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106202
NACRES:
NA.81

product name

(DE3)pLysS Competent Cell Set 2 - Novagen,

biological source

Escherichia coli

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

shipped in

dry ice

storage temp.

−70°C

General description



This product is covered under license from Brookhaven National Labs. Commercial entities need to obtain a research use license prior to purchase. Please contact your licensing department to confirm that your company already holds a research use license.
This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.

Components

•0.2 mlRosetta 2(DE3)pLysS Competent Cells

•0.2 mlRosetta-gami 2(DE3)pLysS Competent Cells

•0.2 mlRosett-gami B(DE3)pLysS Competent Cells

•0.2 mlRosettaBlue(DE3)pLysS Competent Cells

•0.2 mlOrigami 2(DE3)pLysS Competent Cells

•0.2 mlOrigami B(DE3)pLysS Competent Cells

•10 µlTest Plasmid

•4 × 2 mlSOC Medium

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service