Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

70748

Millipore

Pellet Paint® NF Co-Precipitant

Non-fluorescent visible DNA co-precipitant for automated sequencing applications

Synonym(s):

DNA carrier, DNA precipitator, PELLET PAINT® for sequencing, PELLET PAINT® precipitant, RNA carrier, RNA precipitator, non-fluorescent PELLET PAINT®, nucleic acid precipitator, pellet visualizer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500
NACRES:
NA.85
Pricing and availability is not currently available.

Quality Level

form

liquid

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

ambient

storage temp.

−20°C

General description

Note: European Patent No. EP0853680 has been issued to EMD Biosciences, Inc./Novagen for a method for precipitating nucleic acid with a visible carrier. US Patent pending

Pellet Paint NF Co-Precipitant is a nonfluorescent dye-labeled carrier compatible with fluorescent sequencing. It facilitates rapid removal of BigDye Terminators during the alcohol precipitation of cycle sequencingreaction products. Cycle sequencing reactions can be precipitated rapidly with 1 µl of carrierper reaction and centrifugation times of 10minutes. The easily visualized carrier providesa simple confirmation that precipitation has occurred. Sequencing reaction products are efficiently pelleted and dye-labeled terminators remain in the supernatant during alcohol precipitation using the standard Applied Biosystems precipitation protocols. Resuspension of pelleted sequencing reaction products in deionized formamide can beconfirmed by checking for dissolution of the carrier. Pellet Paint NF Co-Precipitantis fully compatible with the ABI PRISM®BigDye Terminator Cycle Sequencing Ready Reaction. To avoid extra sample handling, Pellet Paint NF Co-Precipitant can be added directly to the reaction mix, template DNA, crude PCR samples, or dilution buffer before the cycle sequencing reaction. Although PelletPaint NF absorbs in the UV range, accurate spectrophotometric measurements of DNA or RNA samples are possible; the absorbance ratio (provided with each package of Pellet PaintNF) can be used as a correction factor when determining nucleic acid concentration. PelletPaint NF Co-Precipitant has no detectable effect on the sequencing reaction or sequence accuracy. Pellet Paint NF Co-Precipitant is a useful substitute for the original Pellet Paint Co-Precipitant in applications where fluorescent detection is used.

Features and Benefits

  • Efficient and rapid precipitation of BigDye® cycle sequencing products
  • Efficient removal of dye terminators
  • Direct visualization and tracking of precipitated material
  • No effect on sequencing reaction
  • Substitute for original Pellet Paint Co-Precipitant for fluorescent detection applications

Components

•: 125 µl or 1 ml: Pellet Paint NF Co-Precipitant


•: 1 ml or 8 ml: 3 M Sodium Acetate, pH 5.2

Warning

Toxicity: Standard Handling (A)

Legal Information

This product is covered by U.S. Patents 7,550,447, 7,144,713, EP Patent 853,680, Canadian Patent 2,231,501 owned by EMD Chemicals Inc. or its Affiliates.
ABI Prism is a registered trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
BigDye is a registered trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PELLET PAINT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suyu Quan et al.
Animals : an open access journal from MDPI, 10(2) (2020-02-26)
Milk can mediate maternal-neonatal signal transmission by the bioactive component extracellular vesicles (EVs), which select specific types of miRNA to encapsulate. The miRNA profiling of sheep milk EVs was characterized by sequencing and compared with that of cow milk. Nanoparticle

Questions

  1. Is there a lower limitation of DNA fragment size for use with the Pellet Paint® NF Co-Precipitant 70748? I have DNA fragments at around 500 bp.

    1 answer
    1. there is no recommended lower limitation of DNA fragment size for the Pellet Paint NF Co-Precipitant 70748. However, it notes that precipitation based on ethanol method may lead to lower yield with small DNA fragments, but using DNA fragments at 500 bp should pose no problem.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service