Skip to Content
MilliporeSigma
All Photos(1)

Documents

70562

Sigma-Aldrich

pET-42b(+) DNA - Novagen

Novagen′s pET-41a-c(+) and pET-42a-c(+) vectors incorporate the schistosomal glutathione-S-transferase (GST-Tag) coding sequence as a fusion partner.

Synonym(s):

pET- 41a-c(+) and pET- 42a-c(+) vectors, pET- 42a-c(+) Vector, pET-42 series

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106610
NACRES:
NA.51

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

dry ice
wet ice

storage temp.

−70°C

General description

The pET-41a-c(+) and pET-42a-c(+) vectors incorporate the schistosomal glutathione-S-transferase (GST; GST•Tag) coding sequence as a fusion partner. The GST•Tag sequence has been reported to enhance the production and in some cases the solubility of its fusion partners (Smith 1998). When expressed in a soluble, properly folded form, GST•Tag fusion proteins can be purified with immobilized glutathione. Gentle elution is achieved with buffers containing reduced glutathione. Quantification of soluble GST fusions is also possible by assaying the transferase activity. The pET-41 and -42 series feature the powerful T7lac promoter, and encode the GST•Tag (220 aa) sequence, proteolytic sites, His•Tag (6 aa) sequence, and S•Tag (15 aa) sequence.

In contrast to other commercially available GST-fusion vectors, the Xa (IleGluGlyArg, pET-42 series) and enterokinase (AspAspAspAspLys, pET-41 series) proteolytic cleavage sites have been engineered to allow removal of 100% of the vector-encoded sequences and the generation of native proteins with their authentic N-terminal residues. Unfused proteins can be produced by using the Nde I cloning site. A version of pET-41 is available as a linearized vector prepared for ligation-independent cloning (LIC) of PCR products.

Another pET vector with the GST•Tag sequence is pET-49b(+). Please see the website description for more information. The His•Tag and S•Tag sequences enable alternative protein detection and purification procedures to be performed. For example, when enhancing purity with a separate purification method, or when purifying under denaturing conditions.

The pET-41 series is designed for cloning and high-level expression of peptide sequences fused with the 220 aa GST•Tag protein. Note that the sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map (TB239). The f1 origin is oriented so that infection with helper phage will produce virions containing single stranded DNA that corresponds to the coding strand. Vector encoded sequence can be completely removed when cloning into the PshAI site (as shown below) and then cleaving the GST fusion protein with Enterokinase.

The pET Vectors are supplied as purified plasmid DNA (10 µg). Each order of pET DNA also includes an Induction Control strain (supplied as a glycerol stock). Please contact technical service if you need additional information.

Warning

Toxicity: Standard Handling

Other Notes

Smith, D.B. and Johnson, K.S. 1988. Gene 67, 31.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

This product is sold for internal research use only. Any commercial use of this product, its components, and/or any derivatives thereof (including but not limited to proteins produced using the product or its components) (together and hereinafter the ′EMD Product′) requires signature of a written commercial use agreement with EMD Millipore Corporation or its successor-in-interest. Commercial use shall include but not be limited to: (1) use of the EMD Product to manufacture products for sale to third parties; (2) use of the EMD Product to provide services, information, or data to third parties in exchange for consideration; (3) use of the EMD Product for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the EMD Product, whether or not such EMD Product is resold for research use. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of the EMD Product. Please direct any questions on these use restrictions to: licensing@milliporesigma.com.

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges MilliporeSigma to request certain information about you and the establishment where the GMOs are being handled. Clickhere for Enduser Declaration (EUD) Form.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service