Skip to Content
MilliporeSigma
All Photos(1)

Documents

70189-M

Sigma-Aldrich

pT7Blue Perfectly Blunt® Cloning Kit - Novagen

Synonym(s):

Novagen Blunt Cloning Kits, Perfectly Blunt® Cloning Kits, pT7Blue Vector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106610
NACRES:
NA.54

manufacturer/tradename

Novagen®

Quality Level

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

dry ice

storage temp.

−70°C

General description

The Perfectly Blunt® Cloning Kits are designed for simplified cloning of DNA generated by PCR using any type of DNA polymerase. This approach enables the use of high-fidelity proofreading enzymes for amplification, thus decreasing the probability of generating mutations in the target sequence. In addition, under many conditions blunt cloning is more efficient than T-cloning, most likely due to the observation that the efficiency of single dA addition by Taq DNA polymerase varies significantly depending on the sequence context of the DNA ends, and even the number of PCR cycles performed (Novy 1996, Clark 1998, Brownstein 1996, Magnuson, 1996, Hu 1993).

With the Perfectly Blunt cloning protocol, you can go from PCR product to plating transformants in less than one hour with minimal hands-on time. The finished PCR product is converted to a blunt, phosphorylated form in a 15-minute reaction using premixed reagents. Following a 5-min heat inactivation step, the treated insert is combined with the ready-to-use vector and ligated in an optimized 15-min reaction. An exclusive 8-minute transformation procedure using highly efficient NovaBlue Singles Competent Cells (Cat. No. 70181) generates recombinant colonies that are easily visualized by blue/white screening.

Note that the Perfectly Blunt method is not limited to cloning PCR products; these kits are also suitable for cloning restriction fragments, cDNA, or sheared DNA with the same protocols.

Seven different vectors are available in Perfectly Blunt® Cloning Kits. Vector choices include those designed for general cloning, sequencing, optimal in vitro transcription/translation, and optimal protein expression in E. coli. Each vector is available in a kit containing sufficient reagents for 10, 20, or 40 reactions.

"Vector only” kits are also available in 20- and 40-reaction sizes without ligase and competent cells. For higher-efficiency competent cells, see also pSTBlue-1 Perfectly Blunt Giga Cloning Kit 71229).

The pT7Blue vector features the pUC19 backbone (including high-copy number origin of replication and lac sequences), a T7 promoter, f1 origin of replication, and modified multiple cloning region. The multiple cloning region contains an EcoR V site used for T-cloning flanked by an Nde I site, which allows inserts to be conveniently subcloned into the Nde I site of many pET vectors.

Features and Benefits

  • No restriction enzymes or special primers
  • Compatible with any DNA polymerase
  • Independent of 3ʹ-dA addition or sequence context of DNA ends
  • Blue/white screening with seven different vectors, including pETBlue expression vectors
  • Simple protocol takes less than one hour* from PCR product to plating transformants
* All times listed refer to kits using NovaBlue Singles Competent Cells and ampicillin or chloramphenicol selection. Giga kits, which use NovaBlue GigaSingles Competent Cells, require one hour for outgrowth.

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS

Other Notes

Novy, R.E., et al. 1996. inNovations 6, 7. Clark, J.M. 1988. Nucleic Acids Res. 16, 9677. Brownstein, J.M., et al. 1996. BioTechniques 20, 1004. Magnuson, V.L., et al. 1996. BioTechniques 21, 700. Hu, G. 1993. DNA and Cell Biology 12, 763.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Perfectly Blunt is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service