Skip to Content
MilliporeSigma
All Photos(1)

Documents

70171-M

Sigma-Aldrich

pIE1-neo DNA - Novagen

Novagen′s pIE1-neo plasmid is designed for the establishment of stably-transformed insect cells.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106612
NACRES:
NA.51

Quality Level

form

liquid

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

−20°C

General description

The pIE1-neo plasmid is designed for the establishment of stably-transformed insect cells. When co-transfected with a pIE1 vector containing a foreign gene under the control of the baculovirus ie1 promoter, stable cell lines that constitutively express low levels of the target gene can be selected in the presence of the antibiotic G418 (1). The neo gene is expressed from an "unenhanced" ie1 promoter in pIE1-neo.

Warning

Toxicity: Standard Handling (A)

Other Notes

1. Jarvis, D.L., Weinkauf, C. and Guarino, L.A. (1996) Prot. Exp. Pur. 8, 191–203.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service