Skip to Content
MilliporeSigma
All Photos(1)

Documents

70003-M

Sigma-Aldrich

pBAC-1 DNA - Novagen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106612

form

liquid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

−20°C

General description

pBAC-1 is a baculovirus transfer plasmid designed for simplified cloning and expression of target genes in insect cells. The plasmid is compatible with BacVector-1000, -2000 or -3000 Triple Cut Virus DNA for low background transfection and efficient utilization of the polh promoter. Inserts in pBAC-1 must contain their own translation initiation codon. The vector encodes an optional C-terminal His•Tag fusion sequence that may be utilized if the insert allows read-through in the proper reading frame. The f1 origin is oriented so that infection with helper phage will produce virions containing single stranded DNA that corresponds to the coding strand. Single stranded sequencing of phage-derived DNA can be performed using the 1629DWN primer.

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service