Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69741

Sigma-Aldrich

pET-21b(+) DNA - Novagen

Novagen′s pET-21a-d(+) vectors carry an N-terminal T7-Tag sequence plus an optional C-terminal His-Tag sequence.

Synonym(s):

Novagen DNA Vector, cloning vector, expression vector, pET-21b(+) DNA, pET-21b(+) Plasmid, vector

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$340.00

$340.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$340.00

About This Item

UNSPSC Code:
41106610
NACRES:
NA.51

$340.00


Please contact Customer Service for Availability

Request a Bulk Order

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service