Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

693015

Millipore

Zwittergent® 3-12 Detergent

A synthetic zwitterionic detergent, which unlike other amphoteric surfactants, retains its zwitterionic character over a broad pH range.

Synonym(s):

n-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, hplc solvents

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$212.00
25 G
$754.00
1 KG
$5,690.00

$212.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$212.00
25 G
$754.00
1 KG
$5,690.00

About This Item

Empirical Formula (Hill Notation):
C17H37NO3S
CAS Number:
Molecular Weight:
335.55
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.25

$212.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥99% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

aggregation number

55

color

white

CMC

2 - 4 mM

solubility

water: 5%

shipped in

ambient

storage temp.

15-25°C

SMILES string

[S](=O)(=O)([O-])CCC[N+](CCCCCCCCCCCC)(C)C

InChI

1S/C17H37NO3S/c1-4-5-6-7-8-9-10-11-12-13-15-18(2,3)16-14-17-22(19,20)21/h4-17H2,1-3H3

InChI key

IZWSFJTYBVKZNK-UHFFFAOYSA-N

General description

A synthetic zwitterionic detergent, which unlike other amphoteric surfactants, retains its zwitterionic character over a broad pH range. This property is attributed to the presence of both a strongly basic quaternary ammonium ion and an acidic sulfonate ion of equal strength. This ionic balance presumably prevents irreversible binding to either anionic or cationic compounds.

Application


  • Characterization for the Similarity Assessment between Proposed Biosimilar SB12 and Eculizumab Reference Product Using a State-of-the-Art Analytical Method.: The study characterizes the similarity between biosimilar SB12 and Eculizumab, employing Zwittergent® 3-12 Detergent for accurate protein analysis (Kim et al., 2023).

Warning

Toxicity: Corrosive (L)

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Raper, S.E., et al. 1992. Hepatology16, 433.
Nollstadt, K.H., et al. 1989. J. Chromatogr.497, 87.
Byers, S., et al. 1987. J. Biol. Chem.262, 9166.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Zwittergent is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service