Skip to Content
MilliporeSigma
All Photos(2)

Documents

69036-M

Millipore

Factor Xa, Restriction Grade, Bovine Plasma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

Quality Level

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Find highly-specific cleavage enzymes for isolating proteins during fusion protein purification including enterokinase, factor Xa, HRV 3C protease, and thrombin.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service