Skip to Content
MilliporeSigma
All Photos(1)

Documents

688100

Sigma-Aldrich

YC-1

A nitric oxide-independent, superoxide-sensitive activator of soluble guanylyl cyclase.

Synonym(s):

YC-1, 3-(5ʹ-Hydroxymethyl-2ʹ-furyl)-1-benzylindazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16N2O2
CAS Number:
Molecular Weight:
304.34
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

off-white to pale yellow

solubility

DMSO: 10 mg/mL
methanol: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C19H16N2O2/c22-13-15-10-11-18(23-15)19-16-8-4-5-9-17(16)21(20-19)12-14-6-2-1-3-7-14/h1-11,22H,12-13H2

InChI key

OQQVFCKUDYMWGV-UHFFFAOYSA-N

General description

A nitric oxide-independent, superoxide-sensitive activator of soluble guanylyl cyclase. Inhibits platelet adhesion to collagen and acts as an antithrombotic agent.
A nitric oxide-independent, superoxide-sensitive activator of soluble guanylyl cyclase. Inhibits platelet adhesion to collagen and acts as an antithrombotic agent. Reported to reduce HIF-1α levels and xenograft growth.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Activator of soluble guanylyl cyclase
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). DMSO stock solutions are stable for up to 3 months at -20°C.

Other Notes

Yeo, E.J., et al. 2003. J. Natl Cancer Inst.95, 516.
Mulsch, A., et al. 1997. Br. J. Pharmacol. 120, 681.
Teng, C.M., et al. 1997. Eur. J. Pharmacol. 320, 161.
Wu, C.C., et al. 1997. Biochem. Biophys. Res. Commun. 231, 412.
Wu, C.C., et al. 1995. Br. J. Pharmacol. 116, 1973.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service