Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

681816

Millipore

Lectin, Wheat Germ, Triticum vulgaris

Synonym(s):

WGA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

form

lyophilized solid

Quality Level

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

2-8°C

General description

Specific for tri-N-acetyl-D-glucosamine. Agglutinates erythrocytes and most types of malignant cells more readily than corresponding cells from normal tissues.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and refrigerate (4°C).
Reconstitute at 1 mg/ml in 150 mM NaCl, 10 mM phosphate buffer, pH 7.2-7.4.

Other Notes

Goldstein, I.J., et al. 1975. Biochim. Biophys. Acta405, 53.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service