Skip to Content
MilliporeSigma
All Photos(1)

Documents

681625

Sigma-Aldrich

W-5, Hydrochloride

A cell-permeable and reversible calmodulin antagonist that inhibits myosin light chain kinase (IC₅₀ = 230 µM) and Ca2+-calmodulin-dependent phosphodiesterase (IC₅₀ = 240 µM).

Synonym(s):

W-5, Hydrochloride, N-(6-Aminohexyl)-1-naphthalenesulfonamide, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22N2O2S · xHCl
CAS Number:
Molecular Weight:
306.42 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (NMR)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

DMSO: soluble
water: soluble

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C16H22N2O2S.ClH/c17-12-5-1-2-6-13-18-21(19,20)16-11-7-9-14-8-3-4-10-15(14)16;/h3-4,7-11,18H,1-2,5-6,12-13,17H2;1H

InChI key

HOCSVIGHWPLMFC-UHFFFAOYSA-N

General description

A cell-permeable and reversible calmodulin antagonist that inhibits Ca2+-calmodulin-dependent phosphodiesterase (IC50 = 240 µM) and myosin light chain kinase (IC50 = 230 µM).
A cell-permeable and reversible calmodulin antagonist that inhibits myosin light chain kinase (IC50 = 230 µM) and Ca2+-calmodulin-dependent phosphodiesterase (IC50 = 240 µM).

Biochem/physiol Actions

Cell permeable: yes
Product does not compete with ATP.
Reversible: yes
Target IC50: 230 &microM and 240 µM against myosin light chain kinase and Ca2+-calmodulin-dependent phosphodiesterase, respectively.

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service