Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

681505

Sigma-Aldrich

Anti-CaV2.2 (851-867) Rabbit pAb

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
$517.00

$517.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μL
$517.00

About This Item

UNSPSC Code:
23201100
NACRES:
NA.41

$517.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service