Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

676500-M

Millipore

VEGFR2/Flt3/c-Kit Inhibitor

The VEGFR2/Flt3/c-Kit Inhibitor, also referenced under CAS 796967-10-7, controls the biological activity of VEGFR2/Flt3/c-Kit. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Synonym(s):

VEGFR2/Flt3/c-Kit Inhibitor, N-(4-(3-Amino-1H-indazol-4-yl)phenyl)-Nʹ-(3-methylphenyl)urea, Vascular Endothelial Growth Factor Receptor-2/Flt3/c-Kit Inhibitor, Flt-3 Inhibitor V, VEGFR Tyrosine Kinase Inhibitor XXV, VEGFR2 Kinase Inhibitor XXII, c-Kit Inhibitor I, Vascular Endothelial Growth Factor Receptor-2/Flt3/c-Kit Inhibitor, N-(4-(3-Amino-1H-indazol-4-yl)phenyl)-Nʹ-(3-methylphenyl)urea, Flt-3 Inhibitor V, VEGFR Tyrosine Kinase Inhibitor XXV, VEGFR2 Kinase Inhibitor XXII, c-Kit Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19N5O
CAS Number:
Molecular Weight:
357.41
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

ethanol: 10 mg/mL
DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable 3-aminoindazolylurea compound that acts as a potent and ATP-competitive inhibitor of VEGFR2/KDR (IC50 = 3 nM) as well as Flt3 and cKit (IC50 = 4 to 20 nM). Shown to block VEGF-induced VEGFR2 phosphorylation in 3T3-murine fibroblasts in vitro (IC50 = 13 nM) and exhibit oral availability (AUC = 6.5 µM•h with an oral dose of 10 mg/kg in CD-1 mice) and in vivo efficacy in a murine estradiol-induced uterine edema model (ED50 = 4 mg/kg p.o. in balb/c mice).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Dai, Y., et al. 2007. J. Med. Chem.50, 1584.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service