Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

676490

Sigma-Aldrich

VEGFR2D1-7, Human, Recombinant, S. frugiperda

Synonym(s):

Vascular Endothelial Growth Factor Receptor 2 D1-7, Soluble KDR/Flk-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥90% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

aqueous buffer: ≥100 μg/mL
water: ≥100 μg/mL

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, human VEGFR-2 consisting of all 7 immunoglobulin-like extracellular domains, but lacking the single transmembrane region and the intracellular split tyrosine kinase domain expressed in S. frugiperda insect cells. Contains all of the domains necessary for high affinity binding to VEGF. VEGFR-2 has a lower affinity for VEGF165 than VEGFR-1. However, VEGFR-2 has a higher signaling activity and is reported to mediate almost all observed endothelial cell responses to VEGF, including the increased microvascular permeability and endothelial cell proliferation. VEGFR-2 requires heparin for binding VEGF165.

Biochem/physiol Actions

Measured by the ability to inhibit VEGF165-induced proliferation in human umbilical vein endothelial (HUVE) cells

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 100 mM NaCl, 25 mM MES, pH 5.5.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 6 months at -70°C. Avoid freeze/thaw cycles of solutions.

Other Notes

Robinson, C.J., and Stringer, S.E. 2001 J. Cell Sci.114, 853.
Roeckl, W., et al. 1998. Exp. Cell Res.241, 161.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melis Denizci Öncü et al.
Biotechnology and applied biochemistry, 69(5), 2122-2137 (2021-10-26)
Development of monoclonal antibody therapeutics against vascular endothelial growth factor receptor 2 (VEGFR-2) protein, which is the main regulator in angiogenesis, has been a major challenge for years. In the current study, we engineer an inclusion body forming single-chain variable

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service