Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

672157-M

Sigma-Aldrich

Urokinase Substrate I, Colorimetric - Calbiochem

Synonym(s):

Urokinase Substrate I, Colorimetric - Calbiochem, z-VGR- pNA, z-VGR-pNA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H36N8O7 · xC2H4O2
Molecular Weight:
584.62 (free base basis)
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white to off-white

solubility

water: 5 mg/mL

storage temp.

−20°C

General description

This product has been discontinued.



A synthetic amino acid substrate for urokinase.
Sold on the basis of substrate content.

Biochem/physiol Actions

Primary Target
Urokinase Substrate

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Val-Gly-Arg-pNA

Physical form

Contains 45 mg D-mannitol and 5 mg substrate.

Other Notes

Verheijen, J.H., et al. 1984. Methods Enzym. Anal.5, 425.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service