Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

662140

Sigma-Aldrich

DUB Inhibitor IV, b-AP15

The DUB Inhibitor IV, b-AP15 controls the biological activity of DUB. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

DUB Inhibitor IV, b-AP15, NSC687852, 3,5- bis((4-Nitrophenyl)methylidene)-1-prop-2-enoylpiperidin-4-one, (3 E,5 E)-1-Acryloyl-3,5- bis(4-nitrobenzylidene)piperidin-4-one, USP14 Inhibitor III, UCH-L5/UCH37 Inhibitor II, NSC687852, 3,5-bis((4-Nitrophenyl)methylidene)-1-prop-2-enoylpiperidin-4-one, (3E,5E)-1-Acryloyl-3,5-bis(4-nitrobenzylidene)piperidin-4-one, USP14 Inhibitor III, UCH-L5/UCH37 Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17N3O6
CAS Number:
Molecular Weight:
419.39
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL, clear, yellow

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(/C(CN(C(C=C)=O)C/1)=C/C(C=C2)=CC=C2[N+]([O-])=O)C1=C\C3=CC=C(C=C3)[N+]([O-])=O

InChI

1S/C22H17N3O6/c1-2-21(26)23-13-17(11-15-3-7-19(8-4-15)24(28)29)22(27)18(14-23)12-16-5-9-20(10-6-16)25(30)31/h2-12H,1,13-14H2/b17-11+,18-12+

InChI key

GFARQYQBWJLZMW-JYFOCSDGSA-N

General description

A cell-permeable bis-nitrobenzylidene-piperidinone that inhibits 19S deubiquitinase (DUB) activity (IC50 = 2.1 µM against Ub-AMC hydrolysis) in a reversible manner by directly targeting UCH37/UCH-L5 and USP14, while exhibiting little or no activity toward BAP1, UCH-L1, UCH-L3, USP2, USP7, USP8 (IC50 >100 µM), or POH1/PRN11. Dual UCH37 and USP14 inhibition by a-AP15 results in increased bulk cellular protein ubiquitination level and a blockage of protein proteasomal degradation (Effective conc. ≥770 nM in HCT-116 cells), exactly opposite to the effects seen with USP14 inhibition alone by IU1 (Cat. No. 662210). Effectively suppresses the growth of various tumors in mice (2.5 to 5 mg/kg, i.p. or s.c.) in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

D′Arcy, P., et al. 2011. Nat. Med.17, 1636.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Can any of these DUB inhibitors be used in yeast model system (S. cerevisiae)?

    1 answer
    1. This product has not been tested for suitability in yeast. However, this is not a species specific product. If the 19S deubiquitinase is present in the target organism, the DUB Inhibitor IV, b-AP15 will be effective.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service