Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

657021

Sigma-Aldrich

Tyrosine Kinase Inhibitor Set II

The Tyrosine Kinase Inhibitor Set II controls the biological activity of Tyrosine Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

shipped in

ambient

storage temp.

−20°C

General description

Contains 20 mg of Genistein (Cat. No. 345834), 1 mg of PP2 (Cat. No. 529573), 5 mg of AG 490 (Cat. No. 658401), 5 mg of AG 1296 (Cat. No. 658551), and 5 mg of AG 1478 (Cat. No. 658552). Supplied with an informational insert.
Protein tyrosine phosphorylation is a key element in several membrane signal transduction systems. There are two general classes of tyrosine kinases: the receptor tyrosine kinases and the receptor-associated tyrosine kinases. The receptor tyrosine kinases possess an extracellular ligand binding domain and an intracellular catalytic domain with intrinsic tyrosine kinase activity. The receptor-associated tyrosine kinases transmit signals from the membrane by interacting with the cytoplasmic domain of membrane proteins.



The design of specific inhibitors of tyrosine kinases is important for both fundamental research and for developing therapeutic strategies for the treatment of cancer, atherosclerosis, psoriasis, septic shock, and other disorders where increased tyrosine kinase activity has been reported.



The Tyrosine Kinase Inhibitor Set contains a general tyrosine kinase inhibitor and a series of other inhibitors, which are selective for various tyrosine kinases that are important in cellular signaling. The set contents and characteristics are listed in the table below.

Biochem/physiol Actions

Product competes with ATP.
Secondary Target
PKC, Src family of protein tyrosine kinases, JAK2

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Preparation Note

Concentrated solutions of ≥1 mg/ml should be prepared in anhydrous DMSO.

Other Notes

Hanke, J.H., et al. 1996. J. Biol. Chem.271 695.
Meydan, N., et al. 1996. Nature379, 645.
Levitzi, A., and Gazit, A. 1995. Science267, 1782.
Akiyama, T., et al. 1987. J. Biol. Chem.262, 5592.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service