Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

654256-M

Millipore

TNF-α Inhibitor

The TNF-α Inhibitor, also referenced under CAS 1049741-03-8, controls the biological activity of TNF-α.

Synonym(s):

TNF-α Inhibitor, Tumor Necrosis Factor-α Inhibitor, TNF-α Inhibitor, 6,7-Dimethyl-3-((methyl-(2-(methyl-(1-(3-trifluoromethyl-phenyl)-1H-indol-3-ylmethyl)-amino)-ethyl)-amino)-methyl)-chromen-4-one, diHCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H32F3N3O2 · 2HCl · 0.5H2O
CAS Number:
Molecular Weight:
629.54
MDL number:
UNSPSC Code:
51111800
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 10 mg/mL

storage temp.

−20°C

SMILES string

FC(F)(F)c1cc(ccc1)[n]2c3c(c(c2)CN(CCN(Cc4[c](c5c([o]c4)cc(c(c5)C)C)=O)C)C)cccc3.Cl.Cl

InChI

1S/C32H32F3N3O2.2ClH/c1-21-14-28-30(15-22(21)2)40-20-24(31(28)39)18-37(4)13-12-36(3)17-23-19-38(29-11-6-5-10-27(23)29)26-9-7-8-25(16-26)32(33,34)35;;/h5-11,14-16,19-20H,12-13,17-18H2,1-4H3;2*1H

InChI key

GOZMBJCYMQQACI-UHFFFAOYSA-N

General description

A cell-permeable indolyl-chromenone compound that rapidly inactivates TNF-α by non-covalently binding to the TNF-α trimer and promoting subunit dissociation and preventing TNF-α binding to its receptor (IC50 = 22 µM). Shown to selectively inhibit TNF-α-, but not IL-1β-induced IκB-α degradation in HeLa cells (IC50 = 4.6 µM).

Biochem/physiol Actions

Primary Target
TNF-α
Target IC50: 22 µM for non-covalently binding to the TNF-α trimer and promoting subunit dissociation and preventing TNF-α binding to its receptor

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

He, M.M., et al. 2005. Science310, 1022.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service