Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

654085

Sigma-Aldrich

TTNPB

A synthetic stilbene analog of retinoic acid that acts as a potent retinoic acid receptor (RAR) agonist (EC₅₀ = 21 nM, 4.0 nM, and 2.4 nM for RARα, RARβ, and RARγ, respectively).

Synonym(s):

TTNPB, 4-[(E)-2-(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)-1-propenyl]benzoic Acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$609.00

$609.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$609.00

About This Item

Empirical Formula (Hill Notation):
C24H28O2
CAS Number:
Molecular Weight:
348.48
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$609.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

OC(=O)c1ccc(cc1)\C=C(/C)\c2cc3c(cc2)C(CCC3(C)C)(C)C

InChI

1S/C24H28O2/c1-16(14-17-6-8-18(9-7-17)22(25)26)19-10-11-20-21(15-19)24(4,5)13-12-23(20,2)3/h6-11,14-15H,12-13H2,1-5H3,(H,25,26)/b16-14+

InChI key

FOIVPCKZDPCJJY-JQIJEIRASA-N

General description

A synthetic stilbene analog of retinoic acid which acts as a potent retinoic acid receptor (RAR) agonist (EC50 = 21 nM, 4.0 nM, and 2.4 nM for RARα, RARβ and RARγ, respectively).

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Other Notes

Standeven, A.M., et al. 1996. Toxicol. Appl. Pharmacol. 138, 169.
Beard, R.L., et al. 1995. J.Med. Chem. 38, 2820.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service