Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

65346-M

Sigma-Aldrich

Formalin

Harleco®, suitable for histology, 10%

Synonym(s):

Fixative

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352501
application(s):
diagnostic assay manufacturing
hematology

histology

form

liquid

Quality Level

manufacturer/tradename

Harleco®

concentration

10%

pH

7

mp

-0.1 °C

density

0.98 g/cm3

application(s)

diagnostic assay manufacturing
hematology

histology

storage temp.

10-30°C

General description

HARLECO® Formalin
65346-85 4L Poly Bottle
HARLECO® Neutral Buffered 10%, Recommended for routine use. 10% NBF

Application

HARLECO® Neutral Buffered 10%, Recommended for routine use. 10% NBF
Part of the QSM Histology line. Recommended for routine use.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 1B - Muta. 2 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service