Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

65066-M

Sigma-Aldrich

Hematoxylin Gill II

Harleco® (For Microscopy (contains ethylene glycol, Aluminum sulphate))

Synonym(s):

H&E stain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124

form

liquid

Quality Level

manufacturer/tradename

Harleco® (For Microscopy (contains ethylene glycol, Aluminum sulphate))

mp

-0.1 °C

density

1.22 g/cm3

λmax

545-560 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

HARLECO® Double strength nuclear stain used for progressive or regressive staining in cytology and histology

Application

HARLECO® Double strength nuclear stain used for progressive or regressive staining in cytology and histology
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Same as Gill I but double the strength. May be used for progressive or regressive staining in cytology and histology.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service