Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

648490

Sigma-Aldrich

TRPC3 Channel Inhibitor, Pyr3

The TRPC3 Channel Inhibitor, Pyr3, also referenced under CAS 1160514-60-2, controls the biological activity of TRPC3 Channel. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

TRPC3 Channel Inhibitor, Pyr3, Ethyl-1-(4-(2,3,3-trichloroacrylamide)phenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate, SB-360475, CRAC Channel Inhibitor II

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$210.00

$210.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$210.00

About This Item

Empirical Formula (Hill Notation):
C16H11Cl3F3N3O3
CAS Number:
Molecular Weight:
456.63
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$210.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to beige

solubility

DMSO: 100 mg/mL
ethanol: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

FC(F)(F)c1[n](ncc1C(=O)OCC)c2ccc(cc2)NC(=O)C(=C(Cl)Cl)Cl

InChI

1S/C16H11Cl3F3N3O3/c1-2-28-15(27)10-7-23-25(12(10)16(20,21)22)9-5-3-8(4-6-9)24-14(26)11(17)13(18)19/h3-7H,2H2,1H3,(H,24,26)

InChI key

RZHGONNSASQOAY-UHFFFAOYSA-N

General description

A cell-permeable pyrazole that acts as an equally potent blocker of Stim1 and Orai1-coupled CRAC (Ca2+ release-activated Ca2+) channel-mediated SOCE (store-operated Ca2+ entry) in RBL-2H3 cells (IC50 = 540 nM by Fura-2; peak current density pA/pF = -1.45 and -7.50, respectively, with or without 3 µM BTP2/Pyr3 by whole cell clamp) and the transient receptor potential cation channel TRPC3-mediated ROCE (receptor-operated Ca2+ entry) in TRPC3-expressing HEK293 cells (IC50 = 540 nM; by Fura-2; pA/pF = -2.27 and -18.50, respectively, with or without 3 µM BTP2/Pyr3). Pyr3 is also demonstrated to alleviate pressure overload-induced cardiac hypertrophy following TAC (transverse aortic constriction) operation in mice (0.1 mg/kg/day via i.p. osmotic pump) in vivo. A great complement to Pyr 2 (Cat. No. 203890), Pyr6, and Pyr10 in Ca2+ signaling studies.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Schleifier, H., et al. 2012. Br J PharmacolIn Press
Kiyonaka, S., et al. 2009. Proc. Natl. Acad. Sci. USA106, 5400.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service