Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

648450-M

Millipore

TrkA Inhibitor

The TrkA Inhibitor, also referenced under CAS 388626-12-8, controls the biological activity of TrkA. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

TrkA Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N3O3S
CAS Number:
Molecular Weight:
315.35
UNSPSC Code:
12352200

Assay

≥97% (sum of two isomers, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 200 mg/mL

storage temp.

2-8°C

General description

A potent, cell-permeable, reversible, and highly selective inhibitor of TrkA (IC50 = 63 nM), presumably by targeting the ATP binding pocket. Shown to exhibit ≥150-fold selectivity over Cdk1 and cRaf1.
An oxindole compound that acts as a cell-permeable, reversible, potent, and highly selective inhibitor of TrkA (IC50 = 63 nM), presumably by targeting the kinase’s ATP binding pocket. Shown to exhibit ≥150-fold selectivity over Cdk1 and cRaf1.

Biochem/physiol Actions

Primary Target
TrkA
Target IC50: 63 nM against TrkA

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wood, E.R., et al. 2004. Bioorg. Med. Chem. Lett.14, 953.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service