Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

64245-M

Millipore

L-3,3′,5-Triiodothyronine, Sodium Salt

Stimulates respiratory activity at the mitochondrial level.

Synonym(s):

L-3,3′,5-Triiodothyronine, Sodium Salt, T₃, Na

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11I3NO4 · Na
CAS Number:
Molecular Weight:
672.96
MDL number:
UNSPSC Code:
12352200

Assay

≥95% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

solubility

0.1 M NaOH: 10 mg/mL

storage temp.

10-30°C

InChI

1S/C15H12I3NO4.Na/c16-9-6-8(1-2-13(9)20)23-14-10(17)3-7(4-11(14)18)5-12(19)15(21)22;/h1-4,6,12,20H,5,19H2,(H,21,22);/q;+1/p-1/t12-;/m0./s1

InChI key

SBXXSUDPJJJJLC-YDALLXLXSA-M

General description

Stimulates respiratory activity at the mitochondrial level.

Biochem/physiol Actions

Primary Target
Stimulates respiratory activity at the mitochondrial level

Warning

Toxicity: Harmful (C)

Other Notes

Green, N.K., et al. 1993. Clin. Sci.84, 61.
Lanni, A., et al. 1993. J. Endocrinol.136, 59.
Xu, Q., et al. 1993. Eur. J. Biochem.211, 891.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service