Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

6250

Sigma-Aldrich

Nonfat Powdered Milk

OmniPur® Grade

Synonym(s):

Dehydrated Milk, Nonfat Milk Powder

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$217.00

$217.00


Please contact Customer Service for Availability


Select a Size

Change View
10 G
$217.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.28

$217.00


Please contact Customer Service for Availability

Quality Level

Application

OmniPur® Nonfat Powdered Milk has been used as a blocker:
  • in cell rolling assay[1]
  • to evaluate the interaction of breast cancer circulating tumor cell (CTC) clusters with E-selectin[2]
  • in Western blotting[3]

Packaging

Plastic bag

Legal Information

OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single-cell western blotting
Alex J H, et al.
Nature Methods, 11, 749?755-749?755 (2014)
Siddarth Chandrasekaran et al.
Biomaterials, 33(35), 9037-9048 (2012-09-21)
Hematogenous metastasis involves a glycoprotein mediated adhesion cascade of tumor cells with E-selectin on the endothelial layer of the blood vessels. Cell-cell interactions play a major role in cancer metastasis and invasiveness. Intercellular communication between two cancer cells or between
Michael R King et al.
American journal of physiology. Cell physiology, 308(10), C792-C802 (2015-03-20)
Circulating tumor cells (CTC) have been implicated in the hematogenous spread of cancer. To investigate the fluid phase of cancer from a physical sciences perspective, the multi-institutional Physical Sciences-Oncology Center (PS-OC) Network performed multidisciplinary biophysical studies of single CTC and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service