Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

623

Sigma-Aldrich

Gomori Trichrome Stain

Harleco®, suitable for histology

Synonym(s):

Connective Tissue Stain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121

form

liquid

Quality Level

manufacturer/tradename

Harleco®

λmax

525-535 nm

application(s)

diagnostic assay manufacturing
hematology

histology

shipped in

ambient

storage temp.

15-25°C

General description

623-71 500ml Poly Bottle
HARLECO® Connective tissue stain for collagen and muscle tissue

Application

HARLECO® Connective tissue stain for collagen and muscle tissue
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Connective tissue stain for collagen and muscle tissue.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service