Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

6.18566

Supelco

Heptane-1-sulfonic acid sodium salt monohydrate

for chromatography, LiChropur

Synonym(s):

Heptane-1-sulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NaO3S
CAS Number:
Molecular Weight:
202.25
MDL number:
UNSPSC Code:
12161902
Pricing and availability is not currently available.

grade

for chromatography

product line

LiChropur

SMILES string

[Na+].[S](=O)(=O)([O-])CCCCCCC

InChI

1S/C7H16O3S.Na/c1-2-3-4-5-6-7-11(8,9)10;/h2-7H2,1H3,(H,8,9,10);/q;+1/p-1

InChI key

REFMEZARFCPESH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Development and validation of a reverse phase liquid chromatography method for the simultaneous quantification of eserine and pralidoxime chloride in drugs-in-adhesive matrix type transdermal patches: This study showcases the application of Heptane-1-sulfonic acid sodium salt monohydrate as a crucial ion-pairing agent in HPLC for effective quantification of pharmaceutical compounds. The robust method developed offers a significant advancement in the analysis of complex samples, enhancing both the accuracy and efficiency of pharmaceutical testing and quality control (Banerjee S, Chattopadhyay P, Ghosh A, Kaity S, Veer V, 2013).

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service