Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

6.18506

Supelco

Hexane-1-sulfonic acid sodium salt

for ion pair chromatography, LiChropur

Synonym(s):

Hexane-1-sulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NaO3S
CAS Number:
Molecular Weight:
188.22
MDL number:
UNSPSC Code:
12161904

product line

LiChropur

SMILES string

[Na+].[S](=O)(=O)([O-])CCCCCC

InChI

1S/C6H14O3S.Na/c1-2-3-4-5-6-10(7,8)9;/h2-6H2,1H3,(H,7,8,9);/q;+1/p-1

InChI key

QWSZRRAAFHGKCH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Simultaneous Determination of Sitagliptin Phosphate Monohydrate and Metformin Hydrochloride in Tablets by a Validated UPLC Method.: This study highlights the application of hexane-1-sulfonic acid sodium salt in ultra-performance liquid chromatography (UPLC) for the simultaneous determination of sitagliptin phosphate monohydrate and metformin hydrochloride, enhancing the accuracy and efficiency of pharmaceutical analysis (Malleswararao CS et al., 2012).

  • Simultaneous determination of some water-soluble vitamins and preservatives in multivitamin syrup by validated stability-indicating high-performance liquid chromatography method.: This research utilizes hexane-1-sulfonic acid sodium salt in HPLC to achieve precise quantification of vitamins and preservatives in multivitamin syrups, ensuring product stability and compliance with health standards (Vidović S et al., 2008).

  • Rapid determination of amoxicillin in premixes by HPLC.: Demonstrating the use of hexane-1-sulfonic acid sodium salt for rapid HPLC analysis of amoxicillin in veterinary premixes, this study contributes to enhancing drug safety and regulatory compliance (Dousa M et al., 2005).

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service