Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

6.18215

Sigma-Aldrich

Nickel-aluminium alloy

powder, for the production of Raney nickel for synthesis

Synonym(s):

Nickel-aluminium alloy

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352100

form

powder

InChI

1S/Al.Ni

InChI key

NPXOKRUENSOPAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service