Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

6.18171

Supelco

Hydroxynaphthol blue indicator

Synonym(s):

Hydroxynaphthol blue

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H11N2Na3O11S3
CAS Number:
Molecular Weight:
620.47
UNSPSC Code:
12352107

InChI

1S/C20H14N2O11S3.3Na/c23-15-9-16(35(28,29)30)13-3-1-2-4-14(13)18(15)21-22-19-12-6-5-11(34(25,26)27)7-10(12)8-17(20(19)24)36(31,32)33;;;/h1-9,22,24H,(H,25,26,27)(H,28,29,30)(H,31,32,33);;;/q;3*+1/p-3/b21-18-;;;

InChI key

JQECMNGWLIFQLL-QRQZZYIYSA-K

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service