Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

616376

Sigma-Aldrich

TRAIL, Human, Recombinant, NSO cells

Synonym(s):

Apo2L; TNF-Related Apoptosis-Inducing Ligand

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$638.00

$638.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$638.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$638.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in NSO cells

Quality Level

Assay

≥97% (SDS-PAGE)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1 EU/μg Endotoxin (EU/μg TRAIL)

shipped in

ambient

storage temp.

−70°C

General description

Recombinant, human TRAIL (amino acids 95-281) with an amino-terminal 6X histidine tag and expressed in NSO cells. A type II transmembrane protein with a C-terminal extracellular domain that exhibits homology to other members of this tumor necrosis factor (TNF) family. Displays potent anti-tumor activity against selected targets. Induces apoptosis in a number of transformed cell lines but not in normal tissues. Recombinant bioactive TRAIL is a non-disulfide-linked homotrimer but may contain a small amount (up to 10%) of disulfide-linked homodimers.
Recombinant, human TRAIL expressed in NSO cells a type II transmembrane protein with a C-terminal extracellular domain, which exhibits homology to other TNF family members. Displays potent antitumor activity against selected targets. Induces apoptosis in a number of transformed cell lines but not in normal tissues.

Biochem/physiol Actions

ED₅₀ = 4-12 ng/ml as measured by its ability to induce cytotoxicity in mouse L-929 cell line in the presence of metabolic inhibitors actinomycin D

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from sterile-filtered PBS, 50 µg BSA/µg TRAIL, pH 7.4.

Reconstitution

Following reconstitution, refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C or -70°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or up to 3 months at -20°C or -70°C. Avoid freeze/thaw cycles of solutions.
Reconstitute to ≥20 µg/ml in sterile PBS, 0.1% BSA.

Other Notes

Griffith, T.S., et al. 1999. J. Exp. Med.189, 1343.
Mariani, S.M., et al. 1997. J. Cell Biol.137, 221.
Pan, G., et al. 1997. Science277, 815.
Pan, G., et al. 1997. Science276, 111.
Sheridan, J.P., et al. 1997. Science277, 818.
Pitti, R.M., et al. 1996. J. Biol. Chem.271, 12687.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service